Strontium chloride hexahydrate, tech.
Supplier: Thermo Scientific Chemicals
|
Danger
|
12494-36
12494-A3
CAAA12494-36EA
113.76
CAD
CAAA12494-36
CAAA12494-A3
Strontium chloride hexahydrate, tech.
Strontium chloride hexahydrate
MDL: MFCD00149865
Notes: m.p. 61°C (rapidly heated), 150°C -6H2O
Soluble in water, alcohol
CAS No.: 10025-70-4
Molecular Formula: SrCl2.6H2O
Formula Weight: 266.62 (158.53anhy)
Physical Form: Crystalline
Density: 1.93
Refractive Index: 1.650
Sensitivity: Hygroscopic
Appearance: White to off-white
Solubility: Soluble in water, alcohol
Merck Reference: 14,8840
MDL No.: MFCD00149865
Formula:
SrCl₂·6H₂O Boiling Pt: 100 °C (1013 hPa) Melting Pt: 61 °C Density: 1.93 Storage Temperature: Ambient |
MDL Number:
MFCD00149865 CAS Number: 10025-70-4 EINECS: 233-971-6 Merck Index: 14,8840 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...