Lead(IV) oxide ≥97.0% ACS

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Lead dioxide

12483-22 12483-36
CAAA12483-22EA 115.34 CAD
CAAA12483-22 CAAA12483-36
Lead(IV) oxide ≥97.0% ACS
Lead (IV) oxide

In battery electrodes, manufacturing of pigments and dyes, as analytical lab reagent


Purity: 97.0% min CAS No.: 1309-60-0 Molecular Formula: PbO2 Formula Weight: 239.19 Physical Form: Powder Density: 9.38 Melting Point: 290° dec. Solubility: Insoluble in water. Soluble in HCl (with evolution of chlorine), oxalic acid, HNO3 in presence of H2O2, alkali iodide solutions, hot alkali caustic solutions Merck Reference: 14,5407 UN No.: 1872 MDL No.: MFCD00011165


In battery electrodes, manufacturing of pigments and dyes, as analytical lab reagent

Formula: O₂Pb
MW: 239.2 g/mol
Melting Pt: 290 (dec.)
Density: 9.38
Storage Temperature: Ambient
MDL Number: MFCD00011165
CAS Number: 1309-60-0
EINECS: 215-174-5
UN: 1872
ADR: 5.1 III
Merck Index: 14,5407

Order Now

Specification Test Results

Acid-insoluble matter 0.2%
Carbon compounds (as C) 0.04%
Cl 0.002%
NO3 0.0002
SO3 0.05%
Mn 5 ppm
Ca 0.02%
Cu 0.05%
K 0.05%
NA 0.1%

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR