Copper(II) chloride dihydrate 99+% ACS
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Copper dichloride dihydrate, Cupric chloride dihydrate
12458-22
12458-36
12458-A3
CAAA12458-22EA
95.91
CAD
CAAA12458-22
CAAA12458-36
CAAA12458-A3
Copper(II) chloride dihydrate 99+% ACS
Copper (II) chloride dihydrate
MDL: MFCD00149674
Soluble in water, methanol, ethanol
Purity: 99+%
CAS No.: 10125-13-0
Molecular Formula: CuCl2.2H2O
Formula Weight: 170.48 (134.45anhy)
Physical Form: Granular
Density: 2.510
pH: 3.0-3.8
Melting Point: 100° dec.
Solubility: Soluble in water, methanol, ethanol
Merck Reference: 14,2633
UN No.: 2802
MDL No.: MFCD00149674
Formula:
CuCl₂·2H₂O MW: 170.48 g/mol Melting Pt: 100 °C (dec.) Density: 2.510 Storage Temperature: Ambient |
MDL Number:
MFCD00149674 CAS Number: 10125-13-0 EINECS: 231-210-2 UN: 2802 ADR: 8,III Merck Index: 14,2633 |
Specification Test Results
Insoluble matter | 0.01% |
NO3 | 0.015% |
SO3 | 0.005% |
Ca | 0.005% |
Ni | 0.01% |
K | 0.01% |
NA | 0.0002 |
Fe | 0.00005 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...