Copper(II) chloride, anhydrous ≥98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Isomerization and cracking catalyst, mordant in dyeing and printing fabrics, in electroplating baths for plating Cu on Al, in photography as a fixer, pigment for glass and ceramic
Purity: 98% min
CAS No.: 7447-39-4
Molecular Formula: CuCl2
Formula Weight: 134.45
Physical Form: Powder
Density: 3.386
Appearance: Yellow-brown
Odor: Odorless
pH: 3
Melting Point: 620°
Boiling Point: 993° dec.
Solubility: Soluble in water, alcohol, acetone
Merck Reference: 14,2633
UN No.: 2802
MDL No.: MFCD00010972
Isomerization and cracking catalyst, mordant in dyeing and printing fabrics, in electroplating baths for plating Cu on Al, in photography as a fixer, pigment for glass and ceramic
Formula:
Cl₂Cu Boiling Pt: 993 °C (dec.) Melting Pt: 620 °C Density: 3.386 Storage Temperature: Ambient |
MDL Number:
MFCD00010972 CAS Number: 7447-39-4 EINECS: 231-210-2 UN: 2802 ADR: 8,III Merck Index: 14,2633 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...