Osmium ≥99.8% (metals basis), powder -200 mesh

Supplier: Thermo Scientific Chemicals
Danger

12446-03 12446-06
CAAA12446-03EA 237 CAD
CAAA12446-03 CAAA12446-06
Osmium ≥99.8% (metals basis), powder -200 mesh
Osmium

MDL: MFCD00011147


Purity: 99.8% CAS No.: 7440-04-2 Odor: Strong UN No.: 3089 MDL No.: MFCD00011147

Formula: Os
MW: 190.23 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011147
CAS Number: 7440-04-2
EINECS: 231-114-0
UN: 3089
ADR: 4.1 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR