Yttrium ≥99.9% (REO, rare earth oxide basis), foil, Thickness: 0.025 mm (0.001 in)

Supplier: Thermo Scientific Chemicals
Warning

12443-FF 12443-FI
CAAA12443-FFEA 233.84 CAD
CAAA12443-FF CAAA12443-FI
Yttrium ≥99.9% (REO, rare earth oxide basis), foil, Thickness: 0.025 mm (0.001 in)
Yttrium

MDL: MFCD00011468


Purity: 99.9% CAS No.: 7440-65-5 Physical Form: ≈0.07g/25x25mm, Packaged under argon Odor: Odorless UN No.: 3178 MDL No.: MFCD00011468

Formula: Y
MW: 88.91 g/mol
Boiling Pt: 2927 °C (1013 hPa)
Melting Pt: 1525 °C
Density: 4.472 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011468
CAS Number: 7440-65-5
EINECS: 231-174-8
UN: 3178
ADR: 4.1,III
Merck Index: 12,10240

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR