Yttrium ≥99.6% (REO, rare earth oxide basis), foil, Thickness: 0.1 mm (0.004 in)
Supplier: Thermo Scientific Chemicals
12442-FF
12442-FI
CAAA12442-FFEA
285.98
CAD
CAAA12442-FF
CAAA12442-FI
Yttrium ≥99.6% (REO, rare earth oxide basis), foil, Thickness: 0.1 mm (0.004 in)
Yttrium
Packaged in mylar under argon
Purity: 99.9%
CAS No.: 7440-65-5
Physical Form: ≈0.28g/25x25mm, Packaged in mylar under argon
Odor: Odorless
MDL No.: MFCD00011468
Formula:
Y Boiling Pt: 2927 °C (1013 hPa) Melting Pt: 1525 °C Density: 4.472 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011468 CAS Number: 7440-65-5 EINECS: 231-174-8 Merck Index: 12,10240 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...