Ytterbium ≥99.9% (REO, rare earth oxide basis), foil, Thickness: 0.1 mm (0.004 in)
Supplier: Thermo Scientific Chemicals
|
Danger
|
12437-FF
12437-FI
CAAA12437-FFEA
181.7
CAD
CAAA12437-FF
CAAA12437-FI
Ytterbium ≥99.9% (REO, rare earth oxide basis), foil, Thickness: 0.1 mm (0.004 in)
Ytterbium
MDL: MFCD00011286
Notes: May contain up to 0.15% Calcium
Purity: 99.9%
CAS No.: 7440-64-4
Physical Form: ≈0.43g/25x25mm, Packaged in mylar under argon
Odor: Odorless
UN No.: 3178
MDL No.: MFCD00011286
Formula:
Yb Boiling Pt: 1427 °C (1013 hPa) Melting Pt: 824 °C Density: 6.977 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011286 CAS Number: 7440-64-4 EINECS: 231-173-2 UN: 3178 ADR: 4.1,III Merck Index: 12,10239 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...