Thulium ≥99.9% (REO, rare earth oxide basis), pieces, sublimed dendritic

Supplier: Thermo Scientific Chemicals
Danger

12435-03 12435-06
CAAA12435-03EA 165.9 CAD
CAAA12435-03 CAAA12435-06
Thulium ≥99.9% (REO, rare earth oxide basis), pieces, sublimed dendritic
Thulium

MDL: MFCD00011281


Purity: 99.9% CAS No.: 7440-30-4 Physical Form: Packaged under argon Odor: Odorless UN No.: 3178 MDL No.: MFCD00011281

Formula: Tm
Storage Temperature: Ambient
MDL Number: MFCD00011281
CAS Number: 7440-30-4
EINECS: 231-140-2
UN: 3178
ADR: 4.1,II

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR