Terbium ≥99.9% (REO, rare earth oxide basis), foil, Thickness: 0.1 mm (0.004 in)

Supplier: Thermo Scientific Chemicals
Danger

12430-FF 12430-FI
CAAA12430-FFEA 268.6 CAD
CAAA12430-FF CAAA12430-FI
Terbium ≥99.9% (REO, rare earth oxide basis), foil, Thickness: 0.1 mm (0.004 in)
Terbium

MDL: MFCD00011256


Purity: 99.9% CAS No.: 7440-27-9 Physical Form: ≈0.52g/25x25mm, Packaged in mylar under argon UN No.: 3178 MDL No.: MFCD00011256

Formula: Tb
Boiling Pt: 3041 °C (1013 hPa)
Melting Pt: 1356 °C
Density: 8.27 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011256
CAS Number: 7440-27-9
EINECS: 231-137-6
UN: 3178
ADR: 4.1,II
Merck Index: 12,09300

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR