Scandium ≥99.9% (REO, rare earth oxide basis), foil, Thickness: 0.25 mm (0.01 in)

Supplier: Thermo Scientific Chemicals

12424-FF 12424-FI
CAAA12424-FFEA 750.5 CAD
CAAA12424-FF CAAA12424-FI
Scandium ≥99.9% (REO, rare earth oxide basis), foil, Thickness: 0.25 mm (0.01 in)
Scandium

MDL: MFCD00016323


Purity: 99.9% CAS No.: 7440-20-2 Physical Form: ≈0.5g/25x25mm, Packaged in mylar bag Odor: Odorless MDL No.: MFCD00016323

Formula: Sc
Boiling Pt: 2730 °C (1013 hPa)
Melting Pt: 1541 °C
Density: 2.989 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00016323
CAS Number: 7440-20-2
EINECS: 231-129-2
Merck Index: 12,08537

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR