Samarium ≥99.9% (REO, rare earth oxide basis), pieces, sublimed dendritic

Supplier: Thermo Scientific Chemicals
Danger

12423-09 12423-18 12423-30
CAAA12423-09EA 103.33 CAD
CAAA12423-09 CAAA12423-18 CAAA12423-30
Samarium ≥99.9% (REO, rare earth oxide basis), pieces, sublimed dendritic
Samarium

MDL: MFCD00011233


Purity: 99.9% CAS No.: 7440-19-9 Physical Form: Packaged under argon Appearance: Grey/silvery Odor: Odorless UN No.: 3178 MDL No.: MFCD00011233

Formula: Sm
MW: 150.36 g/mol
Boiling Pt: 1900 °C (1013 hPa)
Melting Pt: 1076.85 °C
Density: 7.42 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011233
CAS Number: 7440-19-9
EINECS: 231-128-7
UN: 3178
ADR: 4.1,III
Merck Index: 12,08498

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR