Lutetium ≥99.9% (REO, rare earth oxide basis), pieces, distilled dendritic

Supplier: Thermo Scientific Chemicals
Danger

12410-03 12410-06
CAAA12410-03EA 404.48 CAD
CAAA12410-03 CAAA12410-06
Lutetium ≥99.9% (REO, rare earth oxide basis), pieces, distilled dendritic
Lutetium

MDL: MFCD00011098


Purity: 99.9% CAS No.: 7439-94-3 Physical Form: Packaged under argon Appearance: Silver Odor: Odorless UN No.: 3178 MDL No.: MFCD00011098

Formula: Lu
Boiling Pt: 3000 °C (1013 hPa)
Melting Pt: 1651…1675 °C
Density: 9.849 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011098
CAS Number: 7439-94-3
EINECS: 231-103-0
UN: 3178
ADR: 4.1,II
Merck Index: 12,05642

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR