Gadolinium ≥99.9% (REO, rare earth oxide basis), foil, thickness 0.025 mm (0.001 in)
Supplier: Thermo Scientific Chemicals
|
Danger
|
12397-FF
12397-FI
CAAA12397-FFEA
267.02
CAD
CAAA12397-FF
CAAA12397-FI
Gadolinium ≥99.9% (REO, rare earth oxide basis), foil, thickness 0.025 mm (0.001 in)
Gadolinium
MDL: MFCD00011022
Purity: 99.9%
CAS No.: 7440-54-2
Physical Form: ≈0.12g/25x25mm, Packaged under argon
Sensitivity: Air and Moisture Sensitive
Appearance: Silver
UN No.: 3178
MDL No.: MFCD00011022
Formula:
Gd Storage Temperature: Ambient |
MDL Number:
MFCD00011022 CAS Number: 7440-54-2 EINECS: 231-162-2 UN: 3178 ADR: 4.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...