Erbium ≥99.9% (REO, rare earth oxide basis), foil, thickness 0.025 mm (0.001 in)
Supplier: Thermo Scientific Chemicals
|
Danger
|
12388-FF
12388-FI
CAAA12388-FFEA
276.5
CAD
CAAA12388-FF
CAAA12388-FI
Erbium ≥99.9% (REO, rare earth oxide basis), foil, thickness 0.025 mm (0.001 in)
Erbium
MDL: MFCD00010987
Purity: 99.9%
CAS No.: 7440-52-0
Physical Form: ≈0.14g/25x25mm, Packaged under argon
UN No.: 3178
MDL No.: MFCD00010987
Formula:
Er MW: 167.26 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00010987 CAS Number: 7440-52-0 EINECS: 231-160-1 UN: 3178 ADR: 4.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...