Erbium ≥99.9% (REO, rare earth oxide basis), foil, thickness 0.25 mm (0.01 in)
Supplier: Thermo Scientific Chemicals
12386-FF
12386-FI
CAAA12386-FFEA
251.22
CAD
CAAA12386-FF
CAAA12386-FI
Erbium ≥99.9% (REO, rare earth oxide basis), foil, thickness 0.25 mm (0.01 in)
Erbium
MDL: MFCD00010987
Purity: 99.9%
CAS No.: 7440-52-0
Physical Form: ≈1.4g/25x25mm
Odor: Odorless
MDL No.: MFCD00010987
Formula:
Er Storage Temperature: Ambient |
MDL Number:
MFCD00010987 CAS Number: 7440-52-0 EINECS: 231-160-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...