Dysprosium ≥99.9% (REO, rare earth oxide basis), foil, thickness 0.025 mm (0.001 in)

Supplier: Thermo Scientific Chemicals
Danger

12383-FF 12383-FI
CAAA12383-FFEA 290.72 CAD
CAAA12383-FF CAAA12383-FI
Dysprosium ≥99.9% (REO, rare earth oxide basis), foil, thickness 0.025 mm (0.001 in)
Dysprosium

MDL: MFCD00010982


Purity: 99.9% CAS No.: 7429-91-6 Physical Form: ≈0.13g/25x25mm, Packaged under argon Appearance: Gray Odor: Odorless UN No.: 3178 MDL No.: MFCD00010982

Formula: Dy
Storage Temperature: Ambient
MDL Number: MFCD00010982
CAS Number: 7429-91-6
EINECS: 231-073-9
UN: 3178
ADR: 4.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR