Dysprosium ≥99.9% (REO, rare earth oxide basis), foil, Thickness: 0.1 mm (0.004 in)

Supplier: Thermo Scientific Chemicals
Danger

12382-FF 12382-FI
CAAA12382-FFEA 271.76 CAD
CAAA12382-FF CAAA12382-FI
Dysprosium ≥99.9% (REO, rare earth oxide basis), foil, Thickness: 0.1 mm (0.004 in)
Dysprosium

MDL: MFCD00010982


Purity: 99.9% CAS No.: 7429-91-6 Physical Form: ≈0.53g/25x25mm, Packaged under argon Appearance: Gray Odor: Odorless UN No.: 3178 MDL No.: MFCD00010982

Formula: Dy
MW: 162.5
Storage Temperature: Ambient
MDL Number: MFCD00010982
CAS Number: 7429-91-6
EINECS: 231-073-9
UN: 3178
ADR: 4.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR