Iron(III) oxide ≥98% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Ferric oxide, Ferric Oxide, (iron(III) oxide), Diiron trioxide
12375-A1
12375-A7
12375-C4
CAAA12375-A1EA
100.33
CAD
CAAA12375-A1
CAAA12375-A7
CAAA12375-C4
Iron(III) oxide ≥98% (metals basis)
Iron (III) oxide
MDL: MFCD00011008
Purity: 98%
CAS No.: 1309-37-1
Molecular Formula: Fe2O3
Formula Weight: 159.69
Physical Form: -325 Mesh Powder
Density: 5.24
Appearance: Red-brown
Odor: Odorless
Melting Point: 1565°
Merck Reference: 14,4028
MDL No.: MFCD00011008
Formula:
Fe₂O₃ MW: 159.69 g/mol Melting Pt: 1565 °C Density: 5.24 Storage Temperature: Ambient |
MDL Number:
MFCD00011008 CAS Number: 1309-37-1 EINECS: 215-168-2 Merck Index: 14,4028 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...