Iron(II,III) oxide ≥97% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Magnetite, Triiron tetraoxide, Iron(II,III) oxide
12374-A1
12374-A7
12374-C4
CAAA12374-A1EA
99.54
CAD
CAAA12374-A1
CAAA12374-A7
CAAA12374-C4
Iron(II,III) oxide ≥97% (metals basis)
Iron (II,III) oxide
MDL: MFCD00011010
Notes: Black powder; may contain a trace amount of Fe2O3 which is red in color
Purity: 97%
CAS No.: 1317-61-9
Molecular Formula: Fe3O4
Formula Weight: 231.54
Physical Form: -325 Mesh Powder
Density: 5.2
Refractive Index: 3.0
Appearance: Black
Odor: Odorless
Melting Point: 1538°
Merck Reference: 14,4040
MDL No.: MFCD00011010
Formula:
Fe₃O₄ MW: 231.54 g/mol Melting Pt: 1538 °C Density: 5.2 Storage Temperature: Ambient |
MDL Number:
MFCD00011010 CAS Number: 1317-61-9 EINECS: 215-277-5 Merck Index: 14,4040 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...