Magnesium hydroxide 95-100.5%

Supplier: Thermo Scientific Chemicals
Warning

12367-22 12367-A4 12367-36
CAAAA12367-22EA 44.08 CAD
CAAAA12367-22 CAAAA12367-A4 CAAA12367-36
Magnesium hydroxide 95-100.5%
Magnesium hydroxide

Flame retardant in plastics, such as PVC, as it produces less toxic fumes


Purity: 95-100.5% CAS No.: 1309-42-8 Molecular Formula: Mg(OH)2 Formula Weight: 58.33 Physical Form: Powder Density: 2.36 Appearance: White Odor: Odorless Melting Point: 360°dec Solubility: Soluble in dilute acids. Practically insoluble in water Merck Reference: 14,5670 MDL No.: MFCD00011104


Flame retardant in plastics, such as PVC, as it produces less toxic fumes

Formula: Mg(OH)₂
Melting Pt: 360 °C (dec.)
Density: 2.36
Storage Temperature: Ambient
MDL Number: MFCD00011104
CAS Number: 1309-42-8
EINECS: 215-170-3
Merck Index: 14,5670

Order Now

Specification Test Results

Form Powder


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR