Aluminium hydroxide ≥76.5%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Hydrargillite, Aluminum hydroxide

12366-22 12366-36 12366-A3
CAAA12366-22EA 40.29 CAD
CAAA12366-22 CAAA12366-36 CAAA12366-A3
Aluminium hydroxide ≥76.5%
Aluminium hydroxide

As absorbent, in chromatography, manufacturing of glass, paper, inks, ceramics, lubricants, cosmetics


Purity: 76.5% min CAS No.: 21645-51-2 Molecular Formula: AlH3O3 Formula Weight: 78.00 Physical Form: Granular Appearance: White Solubility: Practically insoluble in water. Soluble in alkaline aqueous solution, HCl, H2SO4 solution Merck Reference: 14,342 MDL No.: MFCD00003420


As absorbent, in chromatography, manufacturing of glass, paper, inks, ceramics, lubricants, cosmetics

Formula: Al(OH)₃
MW: 78 g/mol
Melting Pt: 300 °C
Density: 2.42 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003420
CAS Number: 21645-51-2
EINECS: 244-492-7
Merck Index: 14,342

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR