Calcium carbonate 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
Marble
Manufacture of paint, rubber, plastics, etc. In food, cosmetics, and pharmaceuticals. As soil conditioner, neutralizer of surface waters, and as an industrial acid neutralizer
Purity: 98%
CAS No.: 471-34-1
Molecular Formula: CaCO3
Formula Weight: 100.09
Physical Form: Powder
Density: 2.930
Refractive Index: 1.6583
Appearance: White
Odor: Odorless
Melting Point: 800° dec.
Solubility: Practically insoluble in water. Soluble in diluted acids
Merck Reference: 14,1657
MDL No.: MFCD00010906
Manufacture of paint, rubber, plastics, etc. In food, cosmetics, and pharmaceuticals. As soil conditioner, neutralizer of surface waters, and as an industrial acid neutralizer
Formula:
CaCO₃ Melting Pt: 800 °C (dec.) Density: 2.930 Storage Temperature: Ambient |
MDL Number:
MFCD00010906 CAS Number: 471-34-1 EINECS: 207-439-9 Merck Index: 14,1657 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...