Calcium nitrate tetrahydrate 99.0-103.0% ACS

Supplier: Thermo Scientific Chemicals
Danger

12364-36 12364-A3
CAAA12364-36EA 90.69 CAD
CAAA12364-36 CAAA12364-A3
Calcium nitrate tetrahydrate 99.0-103.0% ACS
Calcium nitrate tetrahydrate

Manufacturing of other nitrates, fertilizers, matches


Purity: 99.0-103.0% CAS No.: 13477-34-4 Molecular Formula: Ca(NO3)2.4H2O Formula Weight: 236.15 (164.09anhy) Physical Form: Crystalline Density: 1.82 Melting Point: 42° Boiling Point: 132° dec. Solubility: Soluble in water, alcohol, and acetone Merck Reference: 14,1682 UN No.: 1454 MDL No.: MFCD00149604


Manufacturing of other nitrates, fertilizers, matches

Formula: Ca(NO₃)₂·4H₂O
MW: 236.15 g/mol
Boiling Pt: 132 °C (dec.)
Melting Pt: 42 °C
Density: 1.82
Storage Temperature: Ambient
MDL Number: MFCD00149604
CAS Number: 13477-34-4
EINECS: 233-332-1
UN: 1454
ADR: 5.1,III
Merck Index: 14,1682

Order Now

Specification Test Results

Insoluble matter 0.005%
pH of a 5% solution 5.0-7.0 at 25°
Cl 0.005%
NO2 0.001%
SO3 0.002%
Ba 0.005%
Heavy Metals (as Pb) 5 ppm
Fe 5 ppm
MG 0.05%%
K 0.005%%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR