Ammonium nitrate ≥95% ACS

Supplier: Thermo Scientific Chemicals
Danger

12363-36 12363-A3
CAAAA12363-36EA 96.85 CAD
CAAAA12363-36 CAAAA12363-A3
Ammonium nitrate ≥95% ACS
Ammonium nitrate

MDL: MFCD00011425 Common Applications: In fertilizers, making nitrous oxide. Freely soluble in water

Formula: NH₄NO₃
Boiling Pt: 210 °C (dec.)
Melting Pt: 170 °C
Density: 1.72
Storage Temperature: Ambient
MDL Number: MFCD00011425
CAS Number: 6484-52-2
EINECS: 229-347-8
UN: 1942
ADR: 5.1,III
Merck Index: 14,534

Order Now

Specification Test Results

Form Granular
Residue after ignition 0.01%
pH of a 5% solution 4.5-6.0 at 25°
Cl 5 ppm
NO2 Pass test (limit about 5ppm)
PO3 5 ppm
SO3 0.002%
Heavy metals (as Pb) 5 ppm
Fe 2 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR