Aluminium sulfate hydrate 97+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
12362-36
12362-A1
12362-A7
CAAA12362-36EA
75.21
CAD
CAAA12362-36
CAAA12362-A1
CAAA12362-A7
Aluminium sulfate hydrate 97+%
Aluminium sulphate hydrate
MDL: MFCD00149136
Purity: 97+%
CAS No.: 17927-65-0
Molecular Formula: Al2(SO4)3.xH2O (x~14-18)
Formula Weight: 342.15(anhy)
Physical Form: Crystalline
Density: 1.61
pH: 3.5
Merck Reference: 14,366
MDL No.: MFCD00149136
Formula:
Al₂(SO₄)₃·xH₂O Density: 1.61 Storage Temperature: Ambient |
MDL Number:
MFCD00149136 CAS Number: 17927-65-0 EINECS: 233-135-0 Merck Index: 14,366 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...