Ammonium chloride ≥99.5%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Ammonium muriate, Salmiac
As a flux in galvanizing steel sheet, in batteries, to clean soldering irons, analytical lab reagent, in washing powders, electroplating, lustering cotton
Purity: 99.5% min
CAS No.: 12125-02-9
Molecular Formula: NH4Cl
Formula Weight: 53.49
Physical Form: Granular
Density: ≈1.53
Refractive Index: 1.642
Sensitivity: Hygroscopic
Odor: Odorless
Melting Point: 340° subl.
Solubility: Soluble in water, methanol, ethanol. Insoluble in acetone, ether, ethyl acetate.
Merck Reference: 14,509
MDL No.: MFCD00011420
As a flux in galvanizing steel sheet, in batteries, to clean soldering irons, analytical lab reagent, in washing powders, electroplating, lustering cotton
Formula:
NH₄Cl Melting Pt: 340 °C (subl.) Density: 1.53 g/cm³ Storage Temperature: Ambient |
MDL Number:
MFCD00011420 CAS Number: 12125-02-9 EINECS: 235-186-4 Merck Index: 14,509 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...