Aluminium nitrate nonahydrate ≥98%

Supplier: Thermo Scientific Chemicals
Danger

12360-14 12360-30 12360-A1 12360-A7
CAAA12360-14EA 76.47 CAD
CAAA12360-14 CAAA12360-30 CAAA12360-A1 CAAA12360-A7
Aluminium nitrate nonahydrate ≥98%
Aluminium nitrate nonahydrate

MDL: MFCD00003419


Purity: 98% CAS No.: 7784-27-2 Molecular Formula: Al(NO3)3.9H2O Formula Weight: 375.13 (212.99anhy) Physical Form: Crystals Sensitivity: Hygroscopic Merck Reference: 14,352 UN No.: 1438 MDL No.: MFCD00003419

Formula: Al(NO₃)₃·9H₂O
Melting Pt: 73.5 °C
Density: 1.72 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149132
CAS Number: 7784-27-2
EINECS: 236-751-8
UN: 1438
ADR: 5.1,III
Merck Index: 14,352

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR