Nickel (II) oxide ≥99% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Danger
|
12359-18
12359-30
CAAA12359-18EA
80.26
CAD
CAAA12359-18
CAAA12359-30
Nickel (II) oxide ≥99% (metals basis)
Nickel (II) oxide
MDL: MFCD00011145
Purity: 99%
CAS No.: 1313-99-1
Molecular Formula: NiO
Formula Weight: 74.71
Physical Form: -325 Mesh Powder
Density: 6.67
Melting Point: 1984°
Merck Reference: 14,6512
MDL No.: MFCD00011145
Formula:
NiO Melting Pt: 1984 °C Density: 6.67 Storage Temperature: Ambient |
MDL Number:
MFCD00011145 CAS Number: 1313-99-1 EINECS: 215-215-7 Merck Index: 14,6512 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...