Selenium dioxide ≥99.4% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Danger
|
As an oxidizing agent, in manufacturing of other selenium compounds
Purity: 99.4%
CAS No.: 7446-08-4
Molecular Formula: O2Se
Formula Weight: 110.96
Physical Form: Powder
Density: 3.95
Appearance: White to pink
Odor: Odorless
Melting Point: 340-350° subl.
Solubility: Partially soluble in water, methanol, ethanol, acetone. Soluble in concentrated H2SO4
Merck Reference: 14,8434
UN No.: 3283
MDL No.: MFCD00003562
As an oxidizing agent, in manufacturing of other selenium compounds
Formula:
SeO₂ MW: 110.96 g/mol Melting Pt: 315 (subl.) Density: 3.95 Storage Temperature: Ambient |
MDL Number:
MFCD00003562 CAS Number: 7446-08-4 EINECS: 231-194-7 UN: 3283 ADR: 6.1 II Merck Index: 14,8434 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...