Iron(III) chloride, anhydrous 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
MDL: MFCD00011005
Freely soluble in water, alcohol, ether, acetone. Slightly soluble in CS2. Insoluble in ethyl acetate
Purity: 98%
CAS No.: 7705-08-0
Molecular Formula: Cl3Fe
Formula Weight: 162.21
Physical Form: Crystalline
Density: 2.90
Appearance: Black
Odor: Odorless
pH: 1.8
Melting Point: 306°
Boiling Point: 316°
Solubility: Freely soluble in water, alcohol, ether, acetone. Slightly soluble in CS2. Insoluble in ethyl acetate
Merck Reference: 14,4019
UN No.: 1773
MDL No.: MFCD00011005
Formula:
FeCl₃ Boiling Pt: 316 °C Melting Pt: 306 °C Density: 2.90 Storage Temperature: Ambient |
MDL Number:
MFCD00011005 CAS Number: 7705-08-0 EINECS: 231-729-4 UN: 1773 ADR: 8,III Merck Index: 14,4019 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...