Manganese(II) acetate tetrahydrate Mn ∼ 22%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Manganese acetate tetrahydrate, Manganese diacetate tetrahydrate, Acetic acid manganese (II) salt tetrahydrate
Mordant in dyeing, in the manufacture of bister, as a drier for paints and varnishes, and as an oxidation catalyst
CAS No.: 6156-78-1
Molecular Formula: C4H6MnO4.4H2O
Formula Weight: 245.09 (173.03anhy)
Physical Form: Crystalline
Density: 1.589
Appearance: Pale pink
Odor: Pungent
Melting Point: 80°
Solubility: Soluble in water, alcohol
Merck Reference: 14,5724
MDL No.: MFCD00062552
Mordant in dyeing, in the manufacture of bister, as a drier for paints and varnishes, and as an oxidation catalyst
Formula:
(H₃CCOO)₂Mn·4H₂O MW: 245.09 g/mol Melting Pt: 300 Density: 1.589 Storage Temperature: Ambient |
MDL Number:
MFCD00062552 CAS Number: 6156-78-1 EINECS: 211-334-3 Merck Index: 14,5724 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...