Lead(II) chloride 99%, REacton®, Reagent Grade
Supplier: Thermo Scientific Chemicals
|
Danger
|
Analytical reagent, preparation of lead salts, as solder and flux
Purity: 99%
CAS No.: 7758-95-4
Molecular Formula: PbCl2
Formula Weight: 278.10
Physical Form: Crystalline Powder
Density: 5.85
Melting Point: 501°
Boiling Point: 951°
Solubility: Soluble in hot water, alkali hydroxides, NH4Cl solution. Insoluble in cold water and alcohol
Merck Reference: 14,5404
UN No.: 2291
MDL No.: MFCD00011157
Analytical reagent, preparation of lead salts, as solder and flux
Formula:
Cl₂Pb Boiling Pt: 951 °C Melting Pt: 501 °C Density: 5.85 Storage Temperature: Ambient |
MDL Number:
MFCD00011157 CAS Number: 7758-95-4 EINECS: 231-845-5 UN: 2291 ADR: 6.1,III Merck Index: 14,5404 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...