Barium titanate ≥99% (metals basis)

Supplier: Thermo Scientific Chemicals
Warning

12348-18 12348-36 12348-A3
CAAA12348-18EA 63.2 CAD
CAAA12348-18 CAAA12348-36 CAAA12348-A3
Barium titanate ≥99% (metals basis)
Barium titanate

Ferroelectric ceramics, single crystals, storage devices, dielectric amplifiers


Purity: 99% CAS No.: 12047-27-7 Molecular Formula: BaTiO3 Formula Weight: 233.19 Physical Form: -325 Mesh Powder Density: 5.85 Melting Point: 1620° Solubility: Insoluble in water, alkalies. Slightly soluble in dilute acids. Soluble in concentrated acids Merck Reference: 14,1000 MDL No.: MFCD00003447


Ferroelectric ceramics, single crystals, storage devices, dielectric amplifiers

Formula: BaO₃Ti
Melting Pt: 1620 °C
Density: 5.85
Storage Temperature: Ambient
MDL Number: MFCD00003447
CAS Number: 12047-27-7
EINECS: 234-975-0
UN: 1564
ADR: 6.1,III
Merck Index: 14,1000

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR