Barium fluoride ≥99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Ceramic flux, glass manufacture, electronics, and dry-film lubricants
Purity: 99%
CAS No.: 7787-32-8
Molecular Formula: BaF2
Formula Weight: 175.36
Physical Form: -40 Mesh Powder
Density: 4.83
Refractive Index: 1.4741
Odor: Odorless
Melting Point: 1353°
Boiling Point: 2260°
Solubility: Soluble in HCl, HNO3, HF, and aqueous solutions of ammonium chloride
Merck Reference: 14,974
UN No.: 1564
MDL No.: MFCD00003450
Ceramic flux, glass manufacture, electronics, and dry-film lubricants
Formula:
BaF₂ Boiling Pt: 2260 °C Melting Pt: 1353 °C Density: 4.83 Storage Temperature: Ambient |
MDL Number:
MFCD00003450 CAS Number: 7787-32-8 EINECS: 232-108-0 UN: 1564 ADR: 6.1,III Merck Index: 14,974 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...