Tungsten(VI) chloride 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Tungsten hexachloride
12337-06
12337-09
12337-18
12337-30
CAAA12337-06EA
71.42
CAD
CAAA12337-06
CAAA12337-09
CAAA12337-18
CAAA12337-30
Tungsten(VI) chloride 99%
Tungsten (VI) chloride
MDL: MFCD00011463
Purity: 99%
CAS No.: 13283-01-7
Molecular Formula: WCl6
Formula Weight: 396.57
Physical Form: Powder
Density: 3.52
Appearance: Purple
Odor: Pungent
Melting Point: 275°
Boiling Point: 346°
UN No.: 3260
MDL No.: MFCD00011463
Formula:
Cl₆W MW: 396.56 g/mol Boiling Pt: 346 °C Melting Pt: 275 °C Density: 3,52 Storage Temperature: Ambient |
MDL Number:
MFCD00011463 CAS Number: 13283-01-7 EINECS: 236-293-9 UN: 3260 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...