Sodium chloride ≥99.0% ACS
Supplier: Thermo Scientific Chemicals
Warning
|
Synonyms:
Saline, NaCl, Sea salt, Halite
MDL: MFCD00003477
Soluble in water, glycerol. Very slightly soluble in alcohol
Fieser: 6,285
Purity: 99.0% min
CAS No.: 7647-14-5
Molecular Formula: NaCl
Formula Weight: 58.44
Physical Form: Crystalline
Density: 2.165
Refractive Index: 1.5442
Sensitivity: Hygroscopic
Appearance: White
Odor: Odorless
pH: 6-9
Melting Point: 801°
Boiling Point: 1413°
Solubility: Soluble in water, glycerol. Very slightly soluble in alcohol
Merck Reference: 14,8599
MDL No.: MFCD00003477
Formula:
NaCl MW: 58.44 g/mol Boiling Pt: 1413 °C Melting Pt: 801 °C Density: 2.165 Storage Temperature: Ambient |
MDL Number:
MFCD00003477 CAS Number: 7647-14-5 EINECS: 231-598-3 Merck Index: 14,8599 |
Specification Test Results
Insoluble matter | 0.005% |
pH of a 5 % solution | 5.0-9.0 at 25° |
I | 0.002% |
Br | 0.01% |
Chlorate and nitrate (as NO3) | 0.003% |
PO3 | 5 ppm |
SO3 | 0.004% |
Ba | Pass test |
Ca | 0.002%% |
MG | 0.001% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...