Zinc(II) nitrate hexahydrate ≥99% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Danger
|
12313-36
12313-A3
12313-A9
CAAA12313-36EA
113.92
CAD
CAAA12313-36
CAAA12313-A3
CAAA12313-A9
Zinc(II) nitrate hexahydrate ≥99% (metals basis)
Zinc (II) nitrate hexahydrate
MDL: MFCD00149888
Soluble in water, alcohol
Purity: 99%
CAS No.: 10196-18-6
Molecular Formula: N2O6Zn.6H2O
Formula Weight: 297.47 (189.38anhy)
Physical Form: Crystalline
Density: 2.07
Appearance: White
Odor: Odorless
Melting Point: ca 36°
Solubility: Soluble in water, alcohol
Merck Reference: 14,10144
UN No.: 1514
MDL No.: MFCD00149889
Formula:
N₂O₆Zn·H₂O Melting Pt: ∼36 °C Density: 2.07 Storage Temperature: Ambient |
MDL Number:
MFCD00149889 CAS Number: 13778-30-8 EINECS: 231-943-8 UN: 1514 ADR: 5.1,II Merck Index: 14,10144 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...