Calcium chloride dihydrate ≥99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
12312-36
12312-A3
CAAA12312-36EA
127.19
CAD
CAAA12312-36
CAAA12312-A3
Calcium chloride dihydrate ≥99%
Calcium chloride dihydrate
MDL: MFCD00149613
Purity: 99% min
CAS No.: 10035-04-8
Molecular Formula: CaCl2.2H2O
Formula Weight: 147.02 (110.99anhy)
Physical Form: Granular
Density: 0.835
Merck Reference: 14,1659
MDL No.: MFCD00149613
Formula:
CaCl₂·2H₂O Boiling Pt: 1600 °C (1013 hPa) Melting Pt: 176 °C Density: 0.835 Storage Temperature: Ambient |
MDL Number:
MFCD00149613 CAS Number: 10035-04-8 EINECS: 233-140-8 Merck Index: 14,1659 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...