Barium chloride dihydrate ACS

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Barium dichloride

12310-36 12310-A3
CAAA12310-36EA 108.7 CAD
CAAA12310-36 CAAA12310-A3
Barium chloride dihydrate ACS
Barium chloride dihydrate

MDL: MFCD00149154 Very soluble in water. Soluble in methanol. Insoluble in ethanol, acetone, ethyl acetate


CAS No.: 10326-27-9 Molecular Formula: BaCl2.2H2O Formula Weight: 244.28 (208.25anhy) Physical Form: Crystalline Density: 3.097 Odor: Odorless Merck Reference: 14,971 UN No.: 1564 MDL No.: MFCD00149154

Formula: BaCl₂·2H₂O
Boiling Pt: 1560 °C (1013 hPa)
Melting Pt: 113 °C (-2H₂O)
Density: 3.097
Storage Temperature: Ambient
MDL Number: MFCD00149154
CAS Number: 10326-27-9
EINECS: 233-788-1
UN: 1564
ADR: 6.1,III
Merck Index: 14,971

Order Now

Specification Test Results

Insoluble matter 0.005%
Oxidizing substances (as NO3) 0.005%
K 0.0025%
NA 0.005%
Ca 0.05%
Heavy Metals (as Pb) 5 ppm
Fe 2 ppm
Sr 0.001
Loss on drying at 150° 14.0-16.0%
pH of a 5% solution 5.2-8.2 at 25%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR