Zinc chloride 97% ACS
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Zinc dichloride, Dichlorozinc, Zinc (II) chloride
Catalyst, dehydrating and condensing agent, soldering flux, metal etchant
Purity: 97%
CAS No.: 7646-85-7
Molecular Formula: Cl2Zn
Formula Weight: 136.29
Physical Form: Lump
Density: 2.91
Appearance: White
Odor: Odorless
Melting Point: 290°
Boiling Point: 732°
Solubility: Freely soluble in water, dilute HCl, alcohol, glycerol, acetone, ether. Zinc oxychloride forms with much water
Merck Reference: 14,10132
UN No.: 2331
MDL No.: MFCD00011295
Catalyst, dehydrating and condensing agent, soldering flux, metal etchant
Formula:
ZnCl₂ MW: 136.3 g/mol Boiling Pt: 732 °C Melting Pt: 290 °C Density: 2.91 Storage Temperature: Ambient |
MDL Number:
MFCD00011295 CAS Number: 7646-85-7 EINECS: 231-592-0 UN: 2331 ADR: 8 III Merck Index: 14,10132 |
Specification Test Results
Oxychloride | Pass test |
Insoluble matter | 0.005% |
Ca | 0.06% |
MG | 0.01% |
NO3 | 0.003% |
SO3 | 0.01% |
NH4 | 0.005% |
Fe | 0.001% |
PB | 0.005% |
K | 0.02% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...