Cobalt(II) chloride, anhydrous ≥99.7% (metals basis)

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Cobaltous chloride, Cobalt dichloride

12303-18 12303-30
CAAA12303-18EA 129.72 CAD
CAAA12303-18 CAAA12303-30
Cobalt(II) chloride, anhydrous ≥99.7% (metals basis)
Cobalt (II) chloride

MDL: MFCD00010938 . Decomposes at 400°C on long heating in air. Soluble in water, alcohol, acetone, ether, glycerol, and pyridine


Purity: 99.7% CAS No.: 7646-79-9 Molecular Formula: Cl2Co Formula Weight: 129.84 Physical Form: Powder Density: 3.367 Appearance: Pale blue Odor: Odorless Melting Point: 735° Boiling Point: 1049° Solubility: Soluble in water, alcohol, acetone, ether, glycerol, and pyridine Merck Reference: 14,2437 UN No.: 3077 MDL No.: MFCD00010938

Formula: Cl₂Co
MW: 129.84 g/mol
Boiling Pt: 1049
Melting Pt: 735
Density: 3.367
Storage Temperature: Ambient
MDL Number: MFCD00010938
CAS Number: 7646-79-9
EINECS: 231-589-4
UN: 3077
ADR: 9 III
Merck Index: 14,2437

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR