Vanadium(III) oxide ≥95%
Supplier: Thermo Scientific Chemicals
|
Danger
|
12301-30
12301-A1
12301-A7
CAAA12301-30EA
148.36
CAD
CAAA12301-30
CAAA12301-A1
CAAA12301-A7
Vanadium(III) oxide ≥95%
Vanadium (III) oxide
MDL: MFCD00011455
Insoluble in water
Purity: 95%
CAS No.: 1314-34-7
Molecular Formula: O3V2
Formula Weight: 149.88
Physical Form: Powder
Density: 4.87
Appearance: Black
Odor: Odorless
Melting Point: 1970°
Solubility: Insoluble in water
Merck Reference: 14,9923
UN No.: 3285
MDL No.: MFCD00011455
Formula:
O₃V₂ Melting Pt: 1970 °C Density: 4.87 Storage Temperature: Ambient |
MDL Number:
MFCD00011455 CAS Number: 1314-34-7 EINECS: 215-230-9 UN: 3285 ADR: 6.1,II Merck Index: 14,9923 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...