Copper(I) oxide ≥97% (Cu + Cu₂O) (Cu + Cu2O)
Supplier: Thermo Scientific Chemicals
|
Warning
|
Red pigment for glass and ceramic glazes, and as a catalyst
Purity: 97%
CAS No.: 1317-39-1
Molecular Formula: Cu2O
Formula Weight: 143.08
Physical Form: -325 Mesh Powder
Density: 6.0
Refractive Index: 2.705
Sensitivity: Air and Moisture Sensitive
Appearance: Purple-red
Odor: Odorless
Melting Point: 1235°
Boiling Point: 1800° dec.
Solubility: Practically insoluble in water. Soluble in NH4OH
Merck Reference: 14,2664
UN No.: 3077
MDL No.: MFCD00010974
Red pigment for glass and ceramic glazes, and as a catalyst
Formula:
Cu₂O Boiling Pt: 1800 °C (dec.) Melting Pt: 1235 °C Density: 6.0 Storage Temperature: Ambient |
MDL Number:
MFCD00010974 CAS Number: 1317-39-1 EINECS: 215-270-7 UN: 3077 ADR: 9,III Merck Index: 14,2664 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...