Copper(II) oxide 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Cupric oxide, Copper oxide, Copper monoxide granulated

12299-36 12299-A3 12299-A9
CAAA12299-36EA 101.28 CAD
CAAA12299-36 CAAA12299-A3 CAAA12299-A9
Copper(II) oxide 97%
Copper (II) oxide

As a pigment, in sweeting petroleum gases, in galvanic electrodes, metallurgical flux, as an optical glass polishing agent, and as a catalyst in organic reactions


Purity: 97% CAS No.: 1317-38-0 Molecular Formula: CuO Formula Weight: 79.55 Physical Form: -325 Mesh Powder Density: 6.3 Refractive Index: 2.63 Appearance: Black Odor: Odorless Melting Point: 1326° Solubility: Insoluble in water and alcohol. Soluble in dil. acids, NH4Cl and KCN Merck Reference: 14,2646 UN No.: 3077 MDL No.: MFCD00010979


As a pigment, in sweeting petroleum gases, in galvanic electrodes, metallurgical flux, as an optical glass polishing agent, and as a catalyst in organic reactions

Formula: CuO
MW: 79.55 g/mol
Boiling Pt: 2000
Melting Pt: 1326
Density: 6.3
Storage Temperature: Ambient
MDL Number: MFCD00010979
CAS Number: 1317-38-0
EINECS: 215-269-1
UN: 3077
ADR: 9 III
Merck Index: 14,2646

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR