Aluminium chloride hexahydrate, REacton®, Reagent Grade

Supplier: Thermo Scientific Chemicals
Danger

12297-36 12297-A3
CAAA12297-36EA 99.07 CAD
CAAA12297-36 CAAA12297-A3
Aluminium chloride hexahydrate, REacton®, Reagent Grade
Aluminium chloride hexahydrate

Pharmaceuticals, pigments, in cosmetics and deodorants, dyeing fabrics, refining crude oils, parchment paper manufacturing


CAS No.: 7784-13-6 Molecular Formula: AlCl3.6H2O Formula Weight: 241.43 (133.34anhy) Physical Form: Crystalline Density: 2.398 Odor: Odorless pH: 2.5 Melting Point: 100° dec. Solubility: Soluble in water , alcohol, ether, glycerol, propylene glycol Merck Reference: 14,337 UN No.: 3260 MDL No.: MFCD00149134


Pharmaceuticals, pigments, in cosmetics and deodorants, dyeing fabrics, refining crude oils, parchment paper manufacturing

Formula: AlCl₃·6H₂O
Melting Pt: 100 °C (dec.)
Density: 2.398
Storage Temperature: Ambient
MDL Number: MFCD00149134
CAS Number: 7784-13-6
EINECS: 231-208-1
UN: 3260
ADR: 8,III
Merck Index: 14,337

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR