Diboron trioxide ≥97.5% (typ. 98.5%)

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Boron oxide, Boric anhydride

12290-36 12290-A3 12290-A9
CAAA12290-36EA 80.74 CAD
CAAA12290-36 CAAA12290-A3 CAAA12290-A9
Diboron trioxide ≥97.5% (typ. 98.5%)
di-Boron trioxide

As a flux, acid catalyst, or chemical intermediate in production of boron halides, esters, carbide, nitride, and metallic borides


Purity: 97.5% min CAS No.: 1303-86-2 Molecular Formula: B2O3 Formula Weight: 69.62 Physical Form: -60 Mesh Density: 2.46 +/- 0.01 Appearance: White Odor: Odorless Melting Point: ca 450° Boiling Point: 1860° Solubility: Slightly soluble in water @ 25°C. Soluble in water @ 100°C Merck Reference: 14,1337 MDL No.: MFCD00011315


As a flux, acid catalyst, or chemical intermediate in production of boron halides, esters, carbide, nitride, and metallic borides

Formula: B₂O₃
MW: 69.62 g/mol
Boiling Pt: 1860
Melting Pt: 450
Density: 2.46
Storage Temperature: Ambient
MDL Number: MFCD00011315
CAS Number: 1303-86-2
EINECS: 215-125-8
REACH: 01-2119486655-24
Merck Index: 14,1337

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR