Magnesium chloride hexahydrate 98%
Supplier: Thermo Scientific Chemicals
12288-36
12288-A3
12288-A9
CAAA12288-36EA
89.9
CAD
CAAA12288-36
CAAA12288-A3
CAAA12288-A9
Magnesium chloride hexahydrate 98%
Magnesium chloride hexahydrate
MDL: MFCD00149781
Freely soluble in water
Fieser: 10,251
Purity: 98+%
CAS No.: 7791-18-6
Molecular Formula: MgCl2.6H2O
Formula Weight: 203.30 (95.22anhy)
Physical Form: Flake
Density: 1.570
Melting Point: 117° dec.
Merck Reference: 14,5662
MDL No.: MFCD00149781
Formula:
MgCl₂·6H₂O Boiling Pt: 1412 °C (1013 hPa) Melting Pt: 117 °C (dec.) Density: 1.570 Storage Temperature: Ambient |
MDL Number:
MFCD00149781 CAS Number: 7791-18-6 EINECS: 232-094-6 Merck Index: 14,5662 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...