Tellurium tetrachloride ≥99.9% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Danger
|
12284-14
12284-22
CAAA12284-14EA
206.98
CAD
CAAA12284-14
CAAA12284-22
Tellurium tetrachloride ≥99.9% (metals basis)
Tellurium tetrachloride
MDL: MFCD00011262
Decomposes in water forming TeO2 and HCl
Purity: 99.9%
CAS No.: 10026-07-0
Molecular Formula: Cl4Te
Formula Weight: 269.41
Physical Form: Crystalline
Density: 3.01
Appearance: Yellow
Odor: Pungent
Melting Point: 224°
Boiling Point: 380°
Solubility: Decomposes in water forming TeO2 and HCl
Merck Reference: 14,9126
UN No.: 3260
MDL No.: MFCD00011262
Formula:
Cl₄Te Boiling Pt: 380 °C Melting Pt: 224 °C Density: 3.01 Storage Temperature: Ambient |
MDL Number:
MFCD00011262 CAS Number: 10026-07-0 EINECS: 233-055-6 UN: 3260 ADR: 8,II Merck Index: 14,9126 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...