Platinium(IV) oxide monohydrate Pt ≥ 75% (≥75% Pt)

Supplier: Thermo Scientific Chemicals
Danger

12278-03 12278-06
CAAA12278-03EA 417.12 CAD
CAAA12278-03 CAAA12278-06
Platinium(IV) oxide monohydrate Pt ≥ 75% (≥75% Pt)
Platinium (IV) oxide monohydrate

Hydrogenation catalyst. Suitable for the reduction of double and triple bonds, aromatics, carbonyls, nitriles, and nitro groups


CAS No.: 12137-21-2 Molecular Formula: O2Pt.H2O Formula Weight: 245.10 (227.09anhy) Physical Form: Powder, S.A. 85m2/g Appearance: Dark brown to black Melting Point: 450° UN No.: 1479 MDL No.: MFCD00066964


Hydrogenation catalyst. Suitable for the reduction of double and triple bonds, aromatics, carbonyls, nitriles, and nitro groups

Formula: O₂Pt·1H₂O
MW: 245.09 g/mol
Melting Pt: 450 °C
Storage Temperature: Ambient
MDL Number: MFCD00066964
CAS Number: 12137-21-2
EINECS: 215-223-0
UN: 1479
ADR: 5.1,II
Merck Index: 14,7527

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR